siRNA Details
Gene Symbol: BC017158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145590.2 | NP_663565.2 | 7 | 799 | ||
GenBank mRNA | AK148236.1 | - | 7 | 790 | ||
AK149645.1 | - | 7 | 778 | |||
AK154566.1 | - | 7 | 765 | |||
AK164488.1 | - | 7 | 779 | |||
AK165287.1 | - | 7 | 778 | |||
AK170611.1 | - | 7 | 778 | |||
BC017158.1 | - | 7 | 781 | 4919869 | ||
BC024080.1 | - | 6 | 409 | |||
BC094040.1 | - | 7 | 763 |
