siRNA Details
Gene Symbol: BC017158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145590.2 | NP_663565.2 | 2, 3 | 446 | ||
GenBank mRNA | AK148236.1 | - | 2, 3 | 437 | ||
AK149645.1 | - | 2, 3 | 425 | |||
AK154566.1 | - | 2, 3 | 412 | |||
AK164488.1 | - | 2, 3 | 426 | |||
AK165287.1 | - | 2, 3 | 425 | |||
AK170611.1 | - | 2, 3 | 425 | |||
BC017158.1 | - | 2, 3 | 428 | 4919869 | ||
BC024080.1 | - | 1, 2 | 56 | |||
BC094040.1 | - | 2, 3 | 410 |
