siRNA Details
Gene Symbol: Plcg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172285.1 | NP_758489.1 | 20 | 2414 | ||
GenBank mRNA | AK142173.1 | - | 20 | 2417 | ||
AK170430.1 | - | 20 | 2423 | |||
BC019654.1 | - | 7 | 948 | |||
BC023877.1 | - | 20 | 2414 | 5354925 | ||
BC037700.1 | - | 2 | 206 | |||
BC049273.1 | - | 3 | 340 |
