siRNA Details
Gene Symbol: Plcg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172285.1 | NP_758489.1 | 14 | 1472 | ||
GenBank mRNA | AK142173.1 | - | 14 | 1475 | ||
AK151039.1 | - | 14 | 1458 | |||
AK170430.1 | - | 14 | 1476 | |||
BC023877.1 | - | 14 | 1472 | 5354925 |
