siRNA Details
Gene Symbol: Zbtb16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033324.2 | NP_001028496.1 | 5 | 1745 | ||
GenBank mRNA | AK040211.1 | - | 5 | 1738 | ||
AK142947.1 | - | 5 | 1746 | |||
BC132441.1 | - | 4 | 1433 | |||
BC132443.1 | - | 4 | 1433 | |||
BC138775.1 | - | 4 | 1556 | 8861797 | ||
Z47205.1 | - | 5 | 1786 |
