siRNA Details
Gene Symbol: Nyx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173415.4 | NP_775591.1 | 3 | 1567 | ||
GenBank mRNA | AK038912.1 | - | 1 | 961 | ||
AY114303.1 | - | 3267 | ||||
BC035244.1 | - | 3 | 1567 | 5362608 | ||
BC048381.1 | - | 3 | 1566 | 5362608 |
