siRNA Details
Gene Symbol: Slc9a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172780.3 | NP_766368.2 | 16 | 2087 | ||
GenBank mRNA | AK028350.1 | - | 2 | 346 | ||
AK032326.1 | - | 16 | 2068 | |||
AK122232.1 | - | 16 | 2058 | |||
BC130221.1 | - | 16 | 2072 | 40141414 |
