siRNA Details
Gene Symbol: Fancb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146081.1 | NP_001139553.1 | 2 | 748 | ||
NM_175027.4 | NP_778192.3 | 2 | 748 | |||
GenBank mRNA | AK132251.1 | - | 2 | 748 | ||
AK169374.1 | - | 2 | 636 | |||
BC083151.1 | - | 2 | 641 | 30460698 | ||
CJ093028.1 | - | 2 | 636 |
