siRNA Details
Gene Symbol: Ofd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177429.3 | NP_803178.2 | 15 | 1986 | ||
GenBank mRNA | AJ438159.1 | - | 15 | 1837 | ||
BC119070.1 | - | 15 | 1669 | 8733819 | ||
BC120487.1 | - | 15 | 1669 | 40129410 |
