siRNA Details
Gene Symbol: Gas2l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033331.2 | NP_001028503.2 | 9 | 940 | ||
NM_001079876.1 | NP_001073345.1 | 9 | 943 | |||
NM_001284344.1 | NP_001271273.1 | 9 | 940 | |||
GenBank mRNA | AK136054.1 | - | 9 | 953 | ||
AK164505.1 | - | 9 | 929 | |||
AK170634.1 | - | 9 | 875 | |||
AK170750.1 | - | 9 | 865 |
