siRNA Details
Gene Symbol: Gas2l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033331.2 | NP_001028503.2 | 5 | 488 | ||
NM_001079876.1 | NP_001073345.1 | 5 | 491 | |||
NM_001284344.1 | NP_001271273.1 | 5 | 488 | |||
GenBank mRNA | AK136054.1 | - | 5 | 501 | ||
AK164505.1 | - | 5 | 477 | |||
AK170634.1 | - | 5 | 422 | |||
AK170750.1 | - | 5 | 413 | |||
CF165165.1 | - | 5 | 444 |
