siRNA Details
Gene Symbol: Clca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017474.2 | NP_059502.1 | 12 | 2040 | ||
GenBank mRNA | AB017156.1 | - | 12 | 2023 | ||
AK007466.1 | - | 12 | 2038 | |||
AK008659.2 | - | 12 | 2037 | |||
AK033560.1 | - | 2042 | ||||
AK136476.1 | - | 12 | 2037 | |||
BC028343.1 | - | 12 | 2030 | 4217548 | ||
BC116318.2 | - | 12 | 2023 | 40095991 | ||
BC116319.2 | - | 12 | 2023 | 40095992 |
