siRNA Details
Gene Symbol: Zfp58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007575.2 | NP_001007576.1 | 4 | 1403 | ||
GenBank mRNA | AK167399.1 | - | 4 | 1345 | ||
BC027395.1 | - | 1 | 359 | |||
BC139150.1 | - | 4 | 1269 | 8862172 | ||
BC139151.1 | - | 4 | 1269 | 8862173 |
