siRNA Details
Gene Symbol: Pde4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011056.3 | NP_035186.1 | 17 | 2363 | ||
GenBank mRNA | AF536978.1 | - | 17 | 2363 | ||
AK081466.1 | - | 10 | 1736 | |||
AY388962.1 | - | 15 | 1976 | |||
BC132109.1 | - | 16 | 2231 | 40130386 | ||
EU489880.1 | - | 15 | 2219 |
