siRNA Details
Gene Symbol: Zfhx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039198.1 | NP_001034287.1 | 9 | 4631 | ||
GenBank mRNA | AB190186.1 | - | 9 | 4631 |

More Information
Other Species Targeted

Gene Ontology Categories:
