siRNA Details
Gene Symbol: Katna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011835.2 | NP_035965.2 | 4 | 590 | ||
GenBank mRNA | AF153197.1 | - | 4 | 568 | ||
AK012319.1 | - | 4 | 590 | |||
AK041900.1 | - | 4 | 585 | |||
BC009136.1 | - | 4 | 504 | 3597742 |
