siRNA Details
Gene Symbol: Nek4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF099067.1 | - | 3 | 667 | ||
AJ223071.1 | - | 3 | 757 | |||
AK078809.1 | - | 3 | 779 | |||
AK132803.1 | - | 3 | 695 | |||
AK144130.1 | - | 3 | 641 | |||
AK147381.1 | - | 3 | 745 | |||
AK149584.1 | - | 3 | 695 | |||
AK168094.1 | - | 3 | 678 | |||
BC057939.1 | - | 3 | 669 | 5325558 | ||
Y09234.1 | - | 3 | 757 |
