siRNA Details
Gene Symbol: Nek4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF099067.1 | - | 7 | 1468 | ||
AJ223071.1 | - | 7 | 1558 | |||
AK078809.1 | - | 7 | 1580 | |||
AK132803.1 | - | 7 | 1496 | |||
AK149584.1 | - | 7 | 1496 | |||
BC057939.1 | - | 7 | 1470 | 5325558 | ||
CN526970.1 | - | 2 | 464 | |||
Y09234.1 | - | 7 | 1558 |
