siRNA Details
Gene Symbol: Nt5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011851.4 | NP_035981.1 | 7 | 1345 | ||
GenBank mRNA | AK028723.1 | - | 7 | 1315 | ||
AK029979.1 | - | 7 | 1315 | |||
AK128997.1 | - | 7 | 1314 | |||
AK154614.1 | - | 7 | 1322 | |||
BC119268.1 | - | 7 | 1289 | 8734017 | ||
BC119270.1 | - | 7 | 1289 | 8734019 | ||
L12059.1 | - | 7 | 1268 |
