siRNA Details
Gene Symbol: Tenm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290702.1 | NP_001277631.1 | 5 | 1097 | ||
NM_011856.4 | NP_035986.3 | 5 | 1097 | |||
GenBank mRNA | AB025411.1 | - | 5 | 1134 | ||
AK031198.1 | - | 4 | 798 | |||
BC141361.1 | - | 5 | 1098 | 9055973 |
