siRNA Details
Gene Symbol: Dip2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159361.1 | NP_001152833.1 | 12, 13 | 1684 | ||
NM_172819.3 | NP_766407.2 | 7, 8 | 896 | |||
GenBank mRNA | AK030064.1 | - | 7, 8 | 897 | ||
AK147562.1 | - | 12, 13 | 1620 | |||
BC137857.1 | - | 7, 8 | 883 | 8860876 | ||
BC137858.1 | - | 7, 8 | 883 | 8860877 |
