siRNA Details
Gene Symbol: Gmnc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013761.2 | NP_001013783.1 | 3 | 361 | ||
NM_001285916.1 | NP_001272845.1 | 2 | 455 | |||
GenBank mRNA | AK141059.1 | - | 2 | 455 | ||
BC086669.1 | - | 2 | 116 | 30639759 |
