siRNA Details
Gene Symbol: Rasgrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166493.1 | NP_001159965.1 | 2, 3 | 457 | ||
NM_207246.4 | NP_997129.1 | 3, 4 | 449 | |||
GenBank mRNA | AB154824.1 | - | 1, 2 | 144 | ||
AK173041.1 | - | 2, 3 | 342 | |||
BC066069.1 | - | 1, 2 | 87 | 6851971 | ||
BC066127.1 | - | 3, 4 | 374 | 30280332 | ||
CD348455.1 | - | 1, 2 | 97 | |||
CJ131259.1 | - | 3, 4 | 451 | |||
CN695386.1 | - | 2, 3 | 439 |
