siRNA Details
Gene Symbol: 9430020K01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081963.1 | NP_001075432.1 | 3 | 3831 | ||
GenBank mRNA | AK034658.1 | - | 1 | 887 | ||
AK220394.1 | - | 1 | 1988 | |||
BC144919.1 | - | 2 | 3735 | 9053467 |
