siRNA Details
Gene Symbol: Rngtt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF025653.1 | - | 11 | 1178 | ||
AF034568.1 | - | 11 | 1232 | |||
AK002922.1 | - | 11 | 1281 | |||
AK142016.1 | - | 11 | 1275 | |||
AK150497.1 | - | 11 | 1285 | |||
AK151463.1 | - | 11 | 1299 | |||
AK160007.1 | - | 11 | 1287 | |||
AK160706.1 | - | 11 | 1265 | |||
AK169305.1 | - | 11 | 1251 | |||
BC043657.1 | - | 11 | 1250 | 5343539 |
