siRNA Details
Gene Symbol: Rngtt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF025653.1 | - | 7 | 784 | ||
AF034568.1 | - | 7 | 838 | |||
AK002922.1 | - | 7 | 887 | |||
AK079488.1 | - | 7 | 881 | |||
AK142016.1 | - | 7 | 881 | |||
AK150497.1 | - | 7 | 891 | |||
AK151463.1 | - | 7 | 905 | |||
AK160007.1 | - | 7 | 893 | |||
AK160706.1 | - | 7 | 871 | |||
AK169305.1 | - | 7 | 857 | |||
BC043657.1 | - | 7 | 856 | 5343539 |
