siRNA Details
Gene Symbol: Rngtt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF025653.1 | - | 3 | 224 | ||
AF034568.1 | - | 3 | 278 | |||
AK002922.1 | - | 3 | 327 | |||
AK042132.1 | - | 2 | 1637 | |||
AK079488.1 | - | 3 | 324 | |||
AK142016.1 | - | 3 | 321 | |||
AK150497.1 | - | 3 | 331 | |||
AK151463.1 | - | 3 | 345 | |||
AK160007.1 | - | 3 | 333 | |||
AK160706.1 | - | 3 | 311 | |||
AK169305.1 | - | 3 | 297 | |||
BC043657.1 | - | 3 | 296 | 5343539 |
