siRNA Details
Gene Symbol: Malt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172833.2 | NP_766421.1 | 2 | 419 | ||
GenBank mRNA | AK041919.1 | - | 2 | 424 | ||
AK045200.1 | - | 2 | 410 | |||
AK085071.1 | - | 2 | 419 | |||
AK164626.1 | - | 2 | 455 | |||
AK171981.1 | - | 2 | 406 | |||
BC110487.1 | - | 2 | 393 | 40048907 | ||
BC110488.1 | - | 2 | 393 | 40048908 |
