siRNA Details
Gene Symbol: St18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244692.1 | NP_001231621.1 | 12, 13 | 2003 | ||
NM_001244693.1 | NP_001231622.1 | 11, 12 | 1925 | |||
NM_173868.2 | NP_776293.1 | 10, 11 | 1816 | |||
GenBank mRNA | AB097467.1 | - | 10, 11 | 1841 | ||
AJ536155.1 | - | 12, 13 | 1967 | |||
AK032557.1 | - | 12, 13 | 1968 | |||
AK122303.1 | - | 10, 11 | 1765 | |||
AK133743.1 | - | 12, 13 | 1933 | |||
AK137440.1 | - | 10, 11 | 1766 | |||
AK141513.1 | - | 10, 11 | 1725 | |||
AK147320.1 | - | 12, 13 | 2004 | |||
AK147664.1 | - | 12, 13 | 2004 | |||
BC032273.1 | - | 5, 6 | 730 | |||
BC118528.1 | - | 11, 12 | 1839 | 40098452 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_153310, XM_008763490, XM_008763492, XM_008763491, XM_008763494, and others.
Gene Ontology Categories:
