siRNA Details
Gene Symbol: St18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244692.1 | NP_001231621.1 | 7 | 585 | ||
NM_001244693.1 | NP_001231622.1 | 6 | 507 | |||
NM_173868.2 | NP_776293.1 | 5 | 398 | |||
GenBank mRNA | AB097467.1 | - | 5 | 423 | ||
AJ536155.1 | - | 7 | 549 | |||
AK032557.1 | - | 7 | 550 | |||
AK122303.1 | - | 5 | 347 | |||
AK133743.1 | - | 7 | 515 | |||
AK137440.1 | - | 5 | 348 | |||
AK141513.1 | - | 5 | 307 | |||
AK147320.1 | - | 7 | 586 | |||
AK147664.1 | - | 7 | 586 | |||
BC118528.1 | - | 6 | 421 | 40098452 |
