siRNA Details
Gene Symbol: St18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244692.1 | NP_001231621.1 | 12 | 1900 | ||
NM_001244693.1 | NP_001231622.1 | 11 | 1822 | |||
NM_173868.2 | NP_776293.1 | 10 | 1713 | |||
GenBank mRNA | AB097467.1 | - | 10 | 1738 | ||
AJ536155.1 | - | 12 | 1864 | |||
AK032557.1 | - | 12 | 1865 | |||
AK122303.1 | - | 10 | 1662 | |||
AK133743.1 | - | 12 | 1830 | |||
AK141513.1 | - | 10 | 1622 | |||
AK147320.1 | - | 12 | 1901 | |||
AK147664.1 | - | 12 | 1901 | |||
BC032273.1 | - | 5 | 627 | |||
BC118528.1 | - | 11 | 1736 | 40098452 |
