siRNA Details
Gene Symbol: 4933406M09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173771.4 | NP_776132.2 | 4 | 671 | ||
GenBank mRNA | AK030161.1 | - | 4 | 673 | ||
BC116694.1 | - | 3 | 344 | 40105550 | ||
BC116697.1 | - | 3 | 344 | 40105553 |
