siRNA Details
Gene Symbol: Tlk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112705.2 | NP_001106176.1 | 19 | 2370 | ||
NM_001294329.1 | NP_001281258.1 | 17 | 2230 | |||
NM_001294330.1 | NP_001281259.1 | 17 | 2178 | |||
NM_001294331.1 | NP_001281260.1 | 19 | 2383 | |||
NM_001294334.1 | NP_001281263.1 | 19 | 2370 | |||
GenBank mRNA | AF045252.1 | - | 19 | 2389 | ||
AF045253.1 | - | 18 | 1817 | |||
AF045254.1 | - | 14 | 1531 | |||
AK014829.1 | - | 19 | 2008 | |||
AK089925.1 | - | 17 | 1758 | |||
AK161383.1 | - | 1 | 138 | |||
AK165667.1 | - | 19 | 2373 | |||
AX775743.1 | - | 13 | 1395 | |||
BC066198.1 | - | 17 | 1723 | |||
U56909.1 | - | 10 | 1068 |
