siRNA Details
Gene Symbol: Gpr161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081126.1 | NP_001074595.1 | 4 | 1235 | ||
GenBank mRNA | AK145635.1 | - | 3 | 1260 | ||
BC137659.1 | - | 3 | 1173 | 8860678 | ||
EF197953.1 | - | 3 | 1260 |
