siRNA Details
Gene Symbol: Plekhm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039493.1 | NP_001034582.1 | 4 | 1946 | ||
GenBank mRNA | AK034959.1 | - | 4 | 1868 | ||
AK172220.1 | - | 3 | 1039 | |||
BC063098.1 | - | 1946 |
