siRNA Details
Gene Symbol: Neu4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173772.3 | NP_776133.1 | 3 | 576 | ||
GenBank mRNA | AB510404.1 | - | 4 | 557 | ||
AK034236.1 | - | 3 | 547 | |||
AY258421.1 | - | 3 | 489 | |||
BC070438.1 | - | 3 | 576 | 6842155 |
