siRNA Details
Gene Symbol: Zeb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289521.1 | NP_001276450.1 | 7 | 1833 | ||
NM_015753.4 | NP_056568.2 | 8 | 2170 | |||
GenBank mRNA | AF033116.1 | - | 7 | 1685 | ||
AK122312.1 | - | 8 | 1743 | |||
AK141157.1 | - | 7 | 1801 | |||
BC060699.1 | - | 8 | 1907 | 6408845 |
