siRNA Details
Gene Symbol: Iqsec3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033354.3 | NP_001028526.1 | 7 | 2419 | ||
NM_001289685.1 | NP_001276614.1 | 6 | 2148 | |||
GenBank mRNA | AK122446.1 | - | 6 | 2135 | ||
AK147351.1 | - | 6 | 2094 | |||
AK169437.1 | - | 7 | 2419 |
