siRNA Details
Gene Symbol: 2010107G12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025573.2 | NP_001020744.2 | 3 | 414 | ||
GenBank mRNA | BC075651.1 | - | 3 | 414 | 30541436 |

More Information
Other Species Targeted

Gene Ontology Categories:
