siRNA Details
Gene Symbol: E330009J07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175528.4 | NP_780737.1 | 8 | 1160 | ||
GenBank mRNA | AK054277.1 | - | 8 | 1161 | ||
AK122457.1 | - | 7 | 858 | |||
AK139744.1 | - | 2 | 158 | |||
AK147402.1 | - | 8 | 1153 |
