siRNA Details
Gene Symbol: Zfp568 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033355.3 | NP_001028527.2 | 10 | 1969 | ||
NM_001167872.1 | NP_001161344.1 | 10 | 1976 | |||
NM_001167873.1 | NP_001161345.1 | 10 | 1973 | |||
GenBank mRNA | AK143471.1 | - | 10 | 1968 | ||
AK148548.1 | - | 10 | 1976 | |||
BC082606.1 | - | 9 | 1660 | 30672810 |
