siRNA Details
Gene Symbol: Zfp536 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172385.2 | NP_759017.1 | 6 | 2817 | ||
GenBank mRNA | AK030633.1 | - | 6 | 2818 | ||
AK034508.1 | - | 6 | 2761 | |||
AK042625.1 | - | 3 | 877 | |||
AK122270.1 | - | 7 | 2824 | |||
BC033594.1 | - | 6 | 2739 | 5367892 |
