siRNA Details
Gene Symbol: E030002O03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172905.2 | NP_766493.1 | 3 | 341 | ||
GenBank mRNA | AK052956.1 | - | 3 | 357 | ||
AK086819.1 | - | 3 | 356 | |||
BC048933.1 | - | 3 | 322 | |||
BC058791.1 | - | 3 | 341 | 6391535 |
