siRNA Details
Gene Symbol: Ccdc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081429.1 | NP_001074898.1 | 4, 5 | 876 | ||
GenBank mRNA | AK035850.1 | - | 4, 5 | 579 | ||
AK041803.1 | - | 4, 5 | 635 | |||
AK045055.1 | - | 4, 5 | 865 | |||
AK133297.1 | - | 4, 5 | 842 | |||
AK138231.1 | - | 4, 5 | 868 | |||
AK164955.1 | - | 4, 5 | 843 |
