siRNA Details
Gene Symbol: Ccdc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081429.1 | NP_001074898.1 | 7 | 1204 | ||
GenBank mRNA | AK035850.1 | - | 7 | 907 | ||
AK041803.1 | - | 7 | 963 | |||
AK045055.1 | - | 7 | 1193 | |||
AK133297.1 | - | 7 | 1170 | |||
AK138231.1 | - | 7 | 1197 | |||
AK164955.1 | - | 7 | 1171 |
