siRNA Details
Gene Symbol: Usp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003918.2 | NP_001003918.2 | 26 | 2824 | ||
GenBank mRNA | AF548565.1 | - | 26 | 2767 | ||
AK075830.1 | - | 4 | 276 | |||
BC046963.1 | - | 26 | 2762 | |||
BC100666.1 | - | 21 | 2130 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001024790, XM_006245757, XM_006245759, XM_006245756, XM_006245758, and others.
Gene Ontology Categories:
