siRNA Details
Gene Symbol: Usp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003918.2 | NP_001003918.2 | 22 | 2486 | ||
GenBank mRNA | AF548565.1 | - | 22 | 2429 | ||
BC046963.1 | - | 22 | 2424 | |||
BC100666.1 | - | 17 | 1792 |
