siRNA Details
Gene Symbol: Usp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003918.2 | NP_001003918.2 | 8, 9 | 962 | ||
GenBank mRNA | AF548565.1 | - | 8, 9 | 905 | ||
AK135814.1 | - | 1154 | ||||
BC046963.1 | - | 8, 9 | 900 | |||
BC100666.1 | - | 3, 4 | 268 |
