siRNA Details
Gene Symbol: Olfr506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011871.1 | NP_001011871.1 | 1 | 492 | ||
GenBank mRNA | BC134384.1 | - | 496 | |||
BC147670.1 | - | 1 | 521 | 9056643 | ||
BC147675.1 | - | 1 | 521 | 9056648 |
