siRNA Details
Gene Symbol: Olfr282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146457.2 | NP_666668.2 | 1 | 246 | ||
GenBank mRNA | BC128027.1 | - | 1 | 249 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146457.2 | NP_666668.2 | 1 | 246 | ||
GenBank mRNA | BC128027.1 | - | 1 | 249 |